siRNA Details
| Gene Symbol: Csl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027945.3 | NP_082221.2 | 1 | 1051 | ||
| GenBank mRNA | AK005713.1 | - | 1 | 1052 | ||
| AK161295.1 | - | 1 | 857 | |||
| BC050750.1 | - | 1 | 849 | 6775049 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csl| Gene Symbol: Csl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027945.3 | NP_082221.2 | 1 | 1051 | ||
| GenBank mRNA | AK005713.1 | - | 1 | 1052 | ||
| AK161295.1 | - | 1 | 857 | |||
| BC050750.1 | - | 1 | 849 | 6775049 | ||