siRNA Details
| Gene Symbol: 1700003E16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027948.1 | NP_082224.1 | 3 | 485 | ||
| GenBank mRNA | AK005628.1 | - | 2 | 507 | ||
| BC126949.1 | - | 2 | 470 | 40090681 | ||
| BF452482.1 | - | 3 | 387 | |||
| CK137463.1 | - | 2 | 210 | |||

1700003E16Rik
Genomic Map