siRNA Details
| Gene Symbol: Pdia6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027959.3 | NP_082235.1 | 12 | 1245 | ||
| GenBank mRNA | AK076558.1 | - | 12 | 1246 | ||
| AK133792.1 | - | 12 | 1253 | |||
| AK148928.1 | - | 1053 | ||||
| AK151008.1 | - | 12 | 1225 | |||
| AK152960.1 | - | 12 | 1258 | |||
| AK165877.1 | - | 12 | 1224 | |||
| AK166111.1 | - | 12 | 1252 | |||
| AK167386.1 | - | 12 | 1263 | |||
| AK168136.1 | - | 12 | 1252 | |||
| AK168279.1 | - | 12 | 1252 | |||
| BC006865.1 | - | 12 | 1234 | 2645183 | ||

Pdia6
Genomic Map