siRNA Details
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027960.2 | NP_082236.1 | 1 | 321 | ||
| GenBank mRNA | AF488553.1 | - | 1 | 202 | ||
| AK006085.1 | - | 1 | 322 | |||
| BC051148.1 | - | 1 | 331 | 6704315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpep3| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027960.2 | NP_082236.1 | 1 | 321 | ||
| GenBank mRNA | AF488553.1 | - | 1 | 202 | ||
| AK006085.1 | - | 1 | 322 | |||
| BC051148.1 | - | 1 | 331 | 6704315 | ||