siRNA Details
| Gene Symbol: Gpr160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134385.2 | NP_001127857.1 | 4 | 989 | ||
| NM_001134386.2 | NP_001127858.1 | 3 | 472 | |||
| NM_001286994.1 | NP_001273923.1 | 4 | 998 | |||
| NM_001286995.1 | NP_001273924.1 | 3 | 701 | |||
| NM_001286996.1 | NP_001273925.1 | 4 | 572 | |||
| NM_027965.2 | NP_082241.1 | 4 | 801 | |||
| GenBank mRNA | AK006330.1 | - | 4 | 762 | ||
| AK154620.1 | - | 3 | 474 | |||
| AK154784.1 | - | 4 | 956 | |||
| BC116649.1 | - | 4 | 924 | 40088354 | ||

Gpr160
Genomic Map