siRNA Details
| Gene Symbol: Cfap45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027972.1 | NP_082248.1 | 3, 4 | 310 | ||
| GenBank mRNA | AK006449.1 | - | 3, 4 | 302 | ||
| BC150929.1 | - | 3, 4 | 396 | 9087840 | ||
| BC151048.1 | - | 3, 4 | 292 | 9087961 | ||
| BC151055.1 | - | 3, 4 | 292 | 9087968 | ||

Cfap45
Genomic Map