siRNA Details
| Gene Symbol: Cfap45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027972.1 | NP_082248.1 | 5, 6 | 634 | ||
| GenBank mRNA | AK006449.1 | - | 5, 6 | 626 | ||
| BC150929.1 | - | 5, 6 | 720 | 9087840 | ||
| BC151048.1 | - | 5, 6 | 616 | 9087961 | ||
| BC151055.1 | - | 5, 6 | 616 | 9087968 | ||

Cfap45
Genomic Map