siRNA Details
| Gene Symbol: Ppm1j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027982.2 | NP_082258.2 | 8, 9 | 1249 | ||
| GenBank mRNA | AK009235.1 | - | 8, 9 | 1188 | ||
| AY184802.1 | - | 8, 9 | 1249 | |||
| BC117498.1 | - | 8, 9 | 1245 | 40106734 | ||
| BC121788.1 | - | 8, 9 | 1244 | 40106732 | ||

Ppm1j
Genomic Map