siRNA Details
| Gene Symbol: Haus5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027999.1 | NP_082275.1 | 10, 11 | 895 | ||
| GenBank mRNA | AK009476.1 | - | 10, 11 | 875 | ||
| AK129228.1 | - | 7 | 2163 | |||
| AK153920.1 | - | 10, 11 | 877 | |||
| AK163804.1 | - | 10 | 1024 | |||
| BC023723.1 | - | 10, 11 | 845 | |||
| BC089002.1 | - | 10, 11 | 864 | |||
| BC118934.1 | - | 10, 11 | 870 | 40090953 | ||
| BC118935.1 | - | 10, 11 | 870 | 40090954 | ||

Haus5
Genomic Map