siRNA Details
| Gene Symbol: Jsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028001.3 | NP_082277.2 | 4 | 276 | ||
| GenBank mRNA | AK009016.1 | - | 4 | 271 | ||
| AK009578.1 | - | 4 | 491 | |||
| BC104136.1 | - | 3 | 190 | |||
| BC104137.1 | - | 3 | 191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jsrp1| Gene Symbol: Jsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028001.3 | NP_082277.2 | 4 | 276 | ||
| GenBank mRNA | AK009016.1 | - | 4 | 271 | ||
| AK009578.1 | - | 4 | 491 | |||
| BC104136.1 | - | 3 | 190 | |||
| BC104137.1 | - | 3 | 191 | |||