siRNA Details
| Gene Symbol: Zcchc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101433.1 | NP_001094903.1 | 4 | 708 | ||
| GenBank mRNA | BC034735.1 | - | 2 | 262 | ||
| BC147263.1 | - | 4 | 798 | 9056350 | ||
| BC147264.1 | - | 4 | 798 | 9056314 | ||
| BU701574.1 | - | 2 | 196 | |||
| CN526234.1 | - | 4 | 488 | |||

Zcchc24
Genomic Map