siRNA Details
| Gene Symbol: Endod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028013.3 | NP_082289.2 | 2 | 1037 | ||
| GenBank mRNA | AK078056.1 | - | 2 | 788 | ||
| AK079741.1 | - | 2 | 776 | |||
| AK086102.1 | - | 2 | 795 | |||
| AK142446.1 | - | 3 | 715 | |||
| AK173036.1 | - | 2 | 1037 | |||
| AK196557.1 | - | 1 | 257 | |||
| BC019813.1 | - | 2 | 803 | 4017632 | ||
| BC096490.1 | - | 2 | 768 | 6439018 | ||

Endod1
Genomic Map