siRNA Details
| Gene Symbol: 2410016O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023633.3 | NP_076122.2 | 1 | 1835 | ||
| GenBank mRNA | AB041553.1 | - | 1 | 1774 | ||
| AK010521.1 | - | 1 | 941 | |||
| AK080870.1 | - | 1 | 1781 | |||
| BC052152.2 | - | 1 | 1749 | |||
| BC059809.1 | - | 1 | 1786 | |||
| BC138094.1 | - | 1 | 1835 | 8861113 | ||

2410016O06Rik
Genomic Map