siRNA Details
| Gene Symbol: Ist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028018.2 | NP_082294.1 | 2 | 161 | ||
| GenBank mRNA | AK010262.1 | - | 2 | 131 | ||
| AK050468.1 | - | 2 | 128 | |||
| AK088192.1 | - | 2 | 116 | |||
| AK152830.1 | - | 2 | 128 | |||
| AK161899.1 | - | 2 | 117 | |||
| BC017605.1 | - | 2 | 127 | 4507347 | ||
| BC039052.1 | - | 2 | 104 | 2631157 | ||
| BY160969.1 | - | 2 | 162 | |||

Ist1
Genomic Map