siRNA Details
| Gene Symbol: Rnf135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028019.3 | NP_082295.1 | 5 | 1188 | ||
| GenBank mRNA | AK002781.1 | - | 2 | 359 | ||
| AK010429.1 | - | 5 | 1190 | |||
| AK150921.1 | - | 5 | 1189 | |||
| AK152096.1 | - | 5 | 1181 | |||
| BC025209.1 | - | 1 | 351 | |||
| BC096385.1 | - | 5 | 1182 | 30744704 | ||
| BC132307.1 | - | 5 | 1184 | 40130584 | ||
| BC138303.1 | - | 5 | 1184 | 8861322 | ||
| BC145236.1 | - | 4 | 1020 | |||

Rnf135
Genomic Map