siRNA Details
| Gene Symbol: Myh14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271538.1 | NP_001258467.1 | 28, 29 | 4034 | ||
| NM_001271540.1 | NP_001258469.1 | 29, 30 | 3976 | |||
| NM_028021.3 | NP_082297.1 | 27, 28 | 3911 | |||
| GenBank mRNA | AY205605.1 | - | 27, 28 | 3911 | ||
| AY363100.1 | - | 29, 30 | 3976 | |||
| BC041796.1 | - | 2, 3 | 272 | |||
| EF602040.1 | - | 28, 29 | 4034 | |||

Myh14
Genomic Map