siRNA Details
| Gene Symbol: Dnmbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY383729.1 | - | 3 | 764 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnmbp| Gene Symbol: Dnmbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY383729.1 | - | 3 | 764 | ||