siRNA Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077661.1 | NP_001071129.1 | 5, 6 | 695 | ||
| NM_178623.3 | NP_848738.2 | 5, 6 | 279 | |||
| GenBank mRNA | AK033577.1 | - | 5, 6 | 253 | ||
| AK035580.1 | - | 5, 6 | 691 | |||
| AK041953.1 | - | 6, 7 | 944 | |||
| AK129378.1 | - | 4, 5 | 337 | |||
| AK166495.1 | - | 4, 5 | 179 | |||
| AK190812.1 | - | 2, 3 | 68 | |||
| AK211902.1 | - | 1, 2 | 46 | |||
| BC066994.1 | - | 5, 6 | 576 | 5696000 | ||
| BY058620.1 | - | 5, 6 | 279 | |||

Urgcp
Genomic Map