siRNA Details
Gene Symbol: Pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023651.4 | NP_076140.2 | 3 | 986 | ||
GenBank mRNA | AF329877.2 | - | 3 | 895 | ||
AJ304506.1 | - | 3 | 919 | |||
AK011355.1 | - | 3 | 956 | |||
AK029099.1 | - | 3 | 956 | |||
AK029336.1 | - | 3 | 959 | |||
AK031710.1 | - | 3 | 986 | |||
AK031975.1 | - | 3 | 942 | |||
AK032650.1 | - | 2 | 355 | |||
AK153438.1 | - | 3 | 935 | |||
AK169148.1 | - | 3 | 1011 | |||
BC023683.1 | - | 3 | 998 | 5343438 |
