siRNA Details
| Gene Symbol: Tdrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173744.4 | NP_776105.1 | 2, 3 | 339 | ||
| GenBank mRNA | AI506351.1 | - | 2, 3 | 339 | ||
| AK077810.1 | - | 2, 3 | 253 | |||
| AK166875.1 | - | 2, 3 | 250 | |||
| AK166972.1 | - | 2, 3 | 246 | |||
| BC099613.1 | - | 2, 3 | 244 | 30031671 | ||
| BC132465.1 | - | 2, 3 | 252 | 40130742 | ||
| BC132467.1 | - | 2, 3 | 252 | 40130744 | ||
| BC145286.1 | - | 2, 3 | 276 | 9053840 | ||

Tdrp
Genomic Map