siRNA Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028131.3 | NP_082407.1 | 3 | 324 | ||
| GenBank mRNA | AK012097.1 | - | 3 | 325 | ||
| BC027119.1 | - | 1 | 139 | |||
| BC058243.1 | - | 2 | 281 | 6309162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpn| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028131.3 | NP_082407.1 | 3 | 324 | ||
| GenBank mRNA | AK012097.1 | - | 3 | 325 | ||
| BC027119.1 | - | 1 | 139 | |||
| BC058243.1 | - | 2 | 281 | 6309162 | ||