siRNA Details
| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028131.3 | NP_082407.1 | 5, 6 | 449 | ||
| GenBank mRNA | AK012097.1 | - | 5, 6 | 450 | ||
| BC027119.1 | - | 3, 4 | 264 | |||
| BC058243.1 | - | 3, 4 | 360 | 6309162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpn| Gene Symbol: Cenpn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028131.3 | NP_082407.1 | 5, 6 | 449 | ||
| GenBank mRNA | AK012097.1 | - | 5, 6 | 450 | ||
| BC027119.1 | - | 3, 4 | 264 | |||
| BC058243.1 | - | 3, 4 | 360 | 6309162 | ||