siRNA Details
| Gene Symbol: 2510009E07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001881.2 | NP_001001881.1 | 1 | 296 | ||
| GenBank mRNA | AK044207.1 | - | 1 | 214 | ||
| AK051892.1 | - | 1 | 212 | |||
| AK053253.1 | - | 1 | 241 | |||
| BC052490.1 | - | 1 | 251 | |||
| BC072629.1 | - | 1 | 245 | 6833156 | ||
| CJ109106.1 | - | 1 | 296 | |||

2510009E07Rik
Genomic Map