siRNA Details
| Gene Symbol: Fbxl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028149.1 | NP_082425.1 | 11 | 1122 | ||
| GenBank mRNA | AK012109.1 | - | 11 | 846 | ||
| AK144786.1 | - | 11 | 1096 | |||
| AK220232.1 | - | 11 | 1075 | |||
| EF649694.1 | - | 11 | 869 | |||

Fbxl20
Genomic Map