siRNA Details
| Gene Symbol: 1700021F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028158.1 | NP_082434.1 | 1, 2 | 151 | ||
| GenBank mRNA | AK006200.1 | - | 1, 2 | 110 | ||
| BC050790.1 | - | 1, 2 | 143 | 6741974 | ||
| CN835193.1 | - | 1, 2 | 151 | |||

1700021F07Rik
Genomic Map