siRNA Details
| Gene Symbol: 1700030K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028170.2 | NP_082446.2 | 3 | 1069 | ||
| GenBank mRNA | AK089918.1 | - | 3 | 1011 | ||
| AK133815.1 | - | 3 | 1073 | |||
| AK157359.1 | - | 2 | 280 | |||
| AK172202.1 | - | 2 | 278 | |||
| BC008558.1 | - | 3 | 1023 | 3592293 | ||

1700030K09Rik
Genomic Map