siRNA Details
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029911.4 | NP_084187.2 | 7 | 1419 | ||
GenBank mRNA | AK031904.1 | - | 7 | 1350 | ||
AK052327.1 | - | 1 | 282 | |||
AK082153.1 | - | 7 | 1420 | |||
BC132487.1 | - | 7 | 1361 | 40130764 | ||
BC137869.1 | - | 7 | 1361 | 8860888 | ||
DQ185134.1 | - | 7 | 1316 |
