siRNA Details
| Gene Symbol: Smim24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099917.1 | NP_001093387.1 | 2 | 189 | ||
| GenBank mRNA | AK030337.1 | - | 2 | 203 | ||
| BC036156.1 | - | 2 | 193 | |||
| BC036160.1 | - | 2 | 154 | |||
| BC100511.1 | - | 1 | 220 | |||
| BC117023.1 | - | 2 | 189 | 40126342 | ||
| BC117027.1 | - | 2 | 189 | 40126346 | ||

Smim24
Genomic Map