siRNA Details
| Gene Symbol: 1700030F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028180.3 | NP_082456.1 | 3 | 444 | ||
| GenBank mRNA | AK006542.1 | - | 3 | 443 | ||
| BC048613.1 | - | 3 | 444 | 6742336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030F18Rik| Gene Symbol: 1700030F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028180.3 | NP_082456.1 | 3 | 444 | ||
| GenBank mRNA | AK006542.1 | - | 3 | 443 | ||
| BC048613.1 | - | 3 | 444 | 6742336 | ||