siRNA Details
| Gene Symbol: Sh2d4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028182.1 | NP_082458.1 | 6 | 1189 | ||
| GenBank mRNA | AK008803.1 | - | 6 | 975 | ||
| AX776203.1 | - | 5 | 646 | |||
| BC023784.1 | - | 6 | 582 | |||
| BC116682.1 | - | 6 | 897 | 40106612 | ||

Sh2d4a
Genomic Map