siRNA Details
| Gene Symbol: 1810041L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163145.1 | NP_001156617.1 | 4 | 473 | ||
| GenBank mRNA | AK129415.1 | - | 5 | 696 | ||
| BC062953.1 | - | 4 | 473 | 6856297 | ||
| BC119234.1 | - | 4 | 435 | 8733983 | ||
| BC120551.1 | - | 4 | 435 | 40129474 | ||

1810041L15Rik
Genomic Map