siRNA Details
| Gene Symbol: Fryl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028194.2 | NP_082470.2 | 11 | 1354 | ||
| GenBank mRNA | AK033493.1 | - | 11 | 1161 | ||
| AK036035.1 | - | 9 | 1072 | |||
| AK164590.1 | - | 11 | 1355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fryl| Gene Symbol: Fryl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028194.2 | NP_082470.2 | 11 | 1354 | ||
| GenBank mRNA | AK033493.1 | - | 11 | 1161 | ||
| AK036035.1 | - | 9 | 1072 | |||
| AK164590.1 | - | 11 | 1355 | |||