siRNA Details
Gene Symbol: Fryl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028194.2 | NP_082470.2 | 12 | 1490 | ||
GenBank mRNA | AK033493.1 | - | 12 | 1297 | ||
AK036035.1 | - | 10 | 1208 | |||
AK164590.1 | - | 12 | 1491 |
