siRNA Details
| Gene Symbol: Vps9d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028200.1 | NP_082476.1 | 15 | 2000 | ||
| GenBank mRNA | AK005046.2 | - | 15 | 1952 | ||
| AK028751.1 | - | 15 | 1943 | |||
| AK032561.1 | - | 14 | 2457 | |||
| AK151498.1 | - | 14 | 2302 | |||
| AK154409.1 | - | 15 | 1921 | |||
| AK155433.1 | - | 15 | 1921 | |||
| AY751521.1 | - | 15 | 2173 | |||
| BC021813.1 | - | 6 | 803 | |||
| BC057621.1 | - | 15 | 2029 | 6402384 | ||

Vps9d1
Genomic Map