siRNA Details
Gene Symbol: Vps9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028200.1 | NP_082476.1 | 5 | 564 | ||
GenBank mRNA | AK005046.2 | - | 5 | 499 | ||
AK010402.1 | - | 5 | 497 | |||
AK028751.1 | - | 5 | 507 | |||
AK032561.1 | - | 5 | 614 | |||
AK151498.1 | - | 5 | 553 | |||
AK154409.1 | - | 5 | 482 | |||
AK155433.1 | - | 5 | 482 | |||
AY751521.1 | - | 5 | 737 | |||
BC057621.1 | - | 5 | 565 | 6402384 |
