siRNA Details
| Gene Symbol: Usp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033528.1 | NP_001028700.1 | 11 | 1444 | ||
| GenBank mRNA | AK028572.1 | - | 11 | 1489 | ||
| AK049205.1 | - | 11 | 1274 | |||
| AK129363.1 | - | 11 | 1242 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp36| Gene Symbol: Usp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033528.1 | NP_001028700.1 | 11 | 1444 | ||
| GenBank mRNA | AK028572.1 | - | 11 | 1489 | ||
| AK049205.1 | - | 11 | 1274 | |||
| AK129363.1 | - | 11 | 1242 | |||