siRNA Details
| Gene Symbol: Usp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033528.1 | NP_001028700.1 | 13, 14 | 1812 | ||
| GenBank mRNA | AK028572.1 | - | 13, 14 | 1857 | ||
| AK049205.1 | - | 13, 14 | 1642 | |||
| AK129363.1 | - | 13, 14 | 1610 | |||

Usp36
Genomic Map