siRNA Details
| Gene Symbol: Dusp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028207.3 | NP_082483.1 | 3 | 460 | ||
| GenBank mRNA | AF280809.1 | - | 3 | 425 | ||
| AK008734.1 | - | 3 | 441 | |||
| AK079346.1 | - | 3 | 341 | |||
| AK170772.1 | - | 3 | 441 | |||
| AK203252.1 | - | 2 | 196 | |||
| BC016269.1 | - | 3 | 468 | 4489142 | ||

Dusp3
Genomic Map