siRNA Details
| Gene Symbol: Ptar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028208.1 | NP_082484.1 | 2 | 201 | ||
| GenBank mRNA | AK006988.1 | - | 2 | 291 | ||
| AK015225.1 | - | 2 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptar1| Gene Symbol: Ptar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028208.1 | NP_082484.1 | 2 | 201 | ||
| GenBank mRNA | AK006988.1 | - | 2 | 291 | ||
| AK015225.1 | - | 2 | 269 | |||