siRNA Details
| Gene Symbol: Cdpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164625.1 | NP_001158097.1 | 3 | 314 | ||
| NM_001164626.1 | NP_001158098.1 | 2 | 108 | |||
| NM_197998.2 | NP_932115.2 | 3 | 255 | |||
| GenBank mRNA | BC025858.1 | - | 3 | 217 | 5007877 | |
| BQ550379.1 | - | 3 | 273 | |||
| BY081241.1 | - | 3 | 217 | |||

Cdpf1
Genomic Map