siRNA Details
Gene Symbol: Cdpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164625.1 | NP_001158097.1 | 3, 4 | 389 | ||
NM_001164626.1 | NP_001158098.1 | 2, 3 | 183 | |||
NM_197998.2 | NP_932115.2 | 3, 4 | 330 | |||
GenBank mRNA | AK008763.1 | - | 17 | |||
BC025858.1 | - | 3, 4 | 292 | 5007877 | ||
BQ550379.1 | - | 3, 4 | 348 | |||
BY081241.1 | - | 3, 4 | 292 |
