siRNA Details
| Gene Symbol: Psca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028216.2 | NP_082492.1 | 2 | 118 | ||
| GenBank mRNA | AF319173.1 | - | 2 | 106 | ||
| AK008851.1 | - | 2 | 118 | |||
| BC110462.2 | - | 2 | 118 | 40039785 | ||
| EH109693.1 | - | 2 | 115 | |||

Psca
Genomic Map