siRNA Details
| Gene Symbol: 2610028E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011585.1 | - | 3 | 387 | ||
| AK031953.1 | - | 4 | 497 | |||
| AK082619.1 | - | 4 | 422 | |||
| AK083757.1 | - | 5 | 910 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610028E06Rik| Gene Symbol: 2610028E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011585.1 | - | 3 | 387 | ||
| AK031953.1 | - | 4 | 497 | |||
| AK082619.1 | - | 4 | 422 | |||
| AK083757.1 | - | 5 | 910 | |||