siRNA Details
| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028231.2 | NP_082507.1 | 4 | 557 | ||
| GenBank mRNA | AB193520.1 | - | 2 | 167 | ||
| AK012400.1 | - | 4 | 408 | |||
| AK014106.1 | - | 3 | 654 | |||
| AY062429.1 | - | 2 | 167 | |||
| BC046227.1 | - | 4 | 557 | 5703879 | ||
| BC058957.1 | - | 4 | 557 | 5703879 | ||

Kcnmb2
Genomic Map