siRNA Details
| Gene Symbol: Lrpprc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028233.2 | NP_082509.2 | 21 | 2433 | ||
| GenBank mRNA | AK013955.1 | - | 2 | 121 | ||
| BC059862.1 | - | 21 | 2167 | 6417407 | ||
| BC060199.1 | - | 21 | 2205 | |||
| S81177.1 | - | 4 | 349 | |||

Lrpprc
Genomic Map