siRNA Details
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011109.1 | - | 3 | 609 | ||
AK011167.1 | - | 3 | 711 | |||
AK156717.1 | - | 4 | 1032 | |||
AK168313.1 | - | 961 | ||||
BC005726.1 | - | 1 | 135 | |||
BC027368.1 | - | 3 | 916 |
