siRNA Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028248.2 | NP_082524.2 | 15 | 1468 | ||
| GenBank mRNA | AK011959.1 | - | 15 | 1413 | ||
| AK044356.1 | - | 2 | 1631 | |||
| AK050203.1 | - | 15 | 1460 | |||
| AK171811.1 | - | 16 | 1384 | |||
| AK172611.1 | - | 16 | 1384 | |||
| BC031488.1 | - | 15 | 1424 | |||
| CN458042.1 | - | 7 | 603 | |||

Tmem87b
Genomic Map