siRNA Details
| Gene Symbol: Fgfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028263.1 | NP_082539.2 | 2 | 811 | ||
| GenBank mRNA | AK011996.1 | - | 1 | 621 | ||
| AK029452.1 | - | 1 | 614 | |||
| AK167949.1 | - | 2 | 829 | |||
| BC055778.1 | - | 2 | 811 | 6403098 | ||
| DQ503577.1 | - | 1 | 725 | |||

Fgfbp3
Genomic Map