siRNA Details
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175185.4 | NP_780394.1 | 2 | 259 | ||
| GenBank mRNA | AK088412.1 | - | 2 | 243 | ||
| AK162245.1 | - | 2 | 128 | |||
| AK182657.1 | - | 2 | 161 | |||
| BC065074.1 | - | 2 | 175 | 6809862 | ||
| BY282568.1 | - | 2 | 261 | |||

Hsdl1
Genomic Map