siRNA Details
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175185.4 | NP_780394.1 | 3 | 391 | ||
| GenBank mRNA | AK088412.1 | - | 3 | 375 | ||
| AK150638.1 | - | 1 | 58 | |||
| AK152392.1 | - | 1 | 58 | |||
| AK162245.1 | - | 3 | 260 | |||
| BC065074.1 | - | 3 | 307 | 6809862 | ||

Hsdl1
Genomic Map